ENA428683481 Enamine

1-(2,6-dichlorophenyl)cyclobutan-1-amine hydrochloride

  • MDL number MFCD28118522
  • Cas Number : 1803598-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185576

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185576
Availability details for
Quantity
  • Availability
  • Add to Cart