ENA428683482 Enamine

5-[(4-chloro-2-fluorophenyl)methyl]-4-methyl-1,3-thiazol-2-amine hydrobromide

  • MDL number MFCD28118524
  • Cas Number : 1795494-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185587
Availability details for
Quantity
  • Availability
  • Add to Cart