ENA428683486 Enamine

4-(2-fluoro-3-methoxyphenyl)oxan-4-amine hydrochloride

  • MDL number MFCD28118528
  • Cas Number : 1803606-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185595
Availability details for
Quantity
  • Availability
  • Add to Cart