ENA428683487 Enamine

(2S)-1-cyclohexylpropan-2-ol

  • MDL number MFCD28118530
  • Cas Number : 140145-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185637

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185637
Availability details for
Quantity
  • Availability
  • Add to Cart