ENA428683494 Enamine

bis(3-(3-aminopropyl)-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione) trihydrochloride

  • MDL number MFCD28118540
  • Cas Number : 1803566-37-1
  • Molecular Weight : 475.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 1,5HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-185771

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185771
Availability details for
Quantity
  • Availability
  • Add to Cart