ENA428683504 Enamine

1-[3-nitro-5-(trifluoromethyl)phenyl]methanamine hydrochloride

  • MDL number MFCD28118562
  • Cas Number : 1795503-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186763
Availability details for
Quantity
  • Availability
  • Add to Cart