ENA428683505 Enamine

ethyl 5-(1-amino-3-methylbutyl)-1,2,4-oxadiazole-3-carboxylate hydrochloride

  • MDL number MFCD28118563
  • Cas Number : 1803603-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186769
Availability details for
Quantity
  • Availability
  • Add to Cart