ENA428683506 Enamine

[(5-chloro-1,3-dimethyl-1H-pyrazol-4-yl)methyl](methyl)amine dihydrochloride

  • MDL number MFCD28118564
  • Cas Number : 1803590-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186783

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186783
Availability details for
Quantity
  • Availability
  • Add to Cart