ENA428683515 Enamine

2-amino-3-(dimethylamino)-N,N-dimethylbenzene-1-sulfonamide

  • MDL number MFCD28118577
  • Cas Number : 1697594-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186882
Availability details for
Quantity
  • Availability
  • Add to Cart