ENA428683516 Enamine

3-{4-[3-(cyclopropylmethyl)-1,2,4-oxadiazol-5-yl]phenyl}-5-(trifluoromethyl)-1,2,4-oxadiazole

  • MDL number MFCD28118578
  • Cas Number : 1795480-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186918
Availability details for
Quantity
  • Availability
  • Add to Cart