ENA428683520 Enamine

8-(aminomethyl)pentacyclo[5.4.0.0^{2,6}.0^{3,10}.0^{5,9}]undecan-8-ol hydrochloride

  • MDL number MFCD28118582
  • Cas Number : 1795515-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186951
Availability details for
Quantity
  • Availability
  • Add to Cart