ENA428683523 Enamine

[2-amino-2-(furan-2-yl)ethyl]dimethylamine dihydrochloride

  • MDL number MFCD28118585
  • Cas Number : 1803572-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186973
Availability details for
Quantity
  • Availability
  • Add to Cart