ENA428683524 Enamine

1-methyl-5-(propan-2-yl)-1H-pyrazol-4-amine dihydrochloride

  • MDL number MFCD28118587
  • Cas Number : 1803603-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186983

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186983
Availability details for
Quantity
  • Availability
  • Add to Cart