ENA428683527 Enamine

1-{[1,2,4]triazolo[1,5-a]pyrazin-5-yl}piperazine hydrochloride

  • MDL number MFCD28118597
  • Cas Number : 1803591-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187109

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187109
Availability details for
Quantity
  • Availability
  • Add to Cart