ENA428683536 Enamine

2,2,2-trifluoro-1-(1-methyl-1H-pyrazol-4-yl)ethan-1-amine

  • MDL number MFCD11193881
  • Cas Number : 1152551-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185655
Availability details for
Quantity
  • Availability
  • Add to Cart