ENA428683537 Enamine

2-{[(1-methoxypropan-2-yl)oxy]methyl}oxirane

  • MDL number MFCD19636853
  • Cas Number : 132230-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185690

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185690
Availability details for
Quantity
  • Availability
  • Add to Cart