ENA428683541 Enamine

5-ethoxypyrazine-2-carbonitrile

  • MDL number MFCD20284001
  • Cas Number : 1342594-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185699
Availability details for
Quantity
  • Availability
  • Add to Cart