ENA428683543 Enamine

3-(aminomethyl)-1-ethyl-1,2-dihydropyridin-2-one

  • MDL number MFCD19262269
  • Cas Number : 1310149-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185702

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185702
Availability details for
Quantity
  • Availability
  • Add to Cart