ENA428683546 Enamine

5-(4-ethoxyphenyl)-2,3-dihydro-1,3,4-oxadiazol-2-one

  • MDL number MFCD19673357
  • Cas Number : 1343824-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185705
Availability details for
Quantity
  • Availability
  • Add to Cart