ENA428683548 Enamine

5-(3-methoxyphenyl)hexanoic acid

  • MDL number MFCD00666158
  • Cas Number : 121725-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185710
Availability details for
Quantity
  • Availability
  • Add to Cart