ENA428683550 Enamine

5-amino-1-(pyridin-2-yl)-1H-pyrazole-4-carboxamide

  • MDL number MFCD03069409
  • Cas Number : 792953-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185735

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185735
Availability details for
Quantity
  • Availability
  • Add to Cart