ENA428683553 Enamine

3-(pyrazin-2-yloxy)phenol

  • MDL number MFCD12763663
  • Cas Number : 72503-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185738

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185738
Availability details for
Quantity
  • Availability
  • Add to Cart