ENA428683554 Enamine

3-fluoro-2-(piperidin-4-yloxy)pyridine

  • MDL number MFCD16859705
  • Cas Number : 1189578-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185740

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185740
Availability details for
Quantity
  • Availability
  • Add to Cart