ENA428683557 Enamine

5-bromo-3-[(3-propyl-1,2,4-oxadiazol-5-yl)methyl]-3,4-dihydropyrimidin-4-one

  • MDL number MFCD22801443
  • Cas Number : 1515197-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185496
Availability details for
Quantity
  • Availability
  • Add to Cart