ENA428683561 Enamine

3-methoxythiophene-2-carbonyl chloride

  • MDL number MFCD24051494
  • Cas Number : 54450-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185767

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185767
Availability details for
Quantity
  • Availability
  • Add to Cart