ENA428683562 Enamine

4-oxopentanenitrile

  • MDL number MFCD19313148
  • Cas Number : 927-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185500

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P310,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-185500
pictograms GHS06
hstatements H301,H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart