ENA428683563 Enamine

5-bromo-2,3-dihydro-1-benzothiophen-3-amine

  • MDL number MFCD12133516
  • Cas Number : 1156173-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185430
Availability details for
Quantity
  • Availability
  • Add to Cart