ENA428683569 Enamine

3-(2-methyl-1,4-oxazepane-4-carbonyl)-1H-pyrazol-4-amine

  • MDL number MFCD18268270
  • Cas Number : 1341503-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185819

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185819
Availability details for
Quantity
  • Availability
  • Add to Cart