ENA428683571 Enamine

6-fluoro-5-nitroisoquinoline

  • MDL number MFCD27932779
  • Cas Number : 918490-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185506
Availability details for
Quantity
  • Availability
  • Add to Cart