ENA428683577 Enamine

4-methoxy-3-(pyrrolidine-1-sulfonyl)aniline

  • MDL number MFCD09907190
  • Cas Number : 951911-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185887
Availability details for
Quantity
  • Availability
  • Add to Cart