ENA428683579 Enamine

N-(furan-2-ylmethyl)quinazolin-4-amine

  • MDL number MFCD02069202
  • Cas Number : 34116-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185895

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185895
Availability details for
Quantity
  • Availability
  • Add to Cart