ENA428683583 Enamine

1-chloro-2-(difluoromethoxy)-4-methylbenzene

  • MDL number MFCD16303578
  • Cas Number : 1225665-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185922
Availability details for
Quantity
  • Availability
  • Add to Cart