ENA428683584 Enamine

5-amino-N,2-dimethyl-N-(propan-2-yl)benzamide

  • MDL number MFCD14618177
  • Cas Number : 1250353-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185924
Availability details for
Quantity
  • Availability
  • Add to Cart