ENA428683585 Enamine

4-methyl-3-(piperidine-1-carbonyl)aniline

  • MDL number MFCD14618181
  • Cas Number : 1251326-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185925
Availability details for
Quantity
  • Availability
  • Add to Cart