ENA428683588 Enamine

5-bromo-7-methyl-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD18647722
  • Cas Number : 1273609-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185931
Availability details for
Quantity
  • Availability
  • Add to Cart