ENA428683591 Enamine

2-(3-methoxyphenyl)-2-oxoethyl 6-cyclopropyl-3-methyl-[1,2]oxazolo[5,4-b]pyridine-4-carboxylate

  • MDL number MFCD28118553
  • Cas Number : 1788977-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185935

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185935
Availability details for
Quantity
  • Availability
  • Add to Cart