ENA428683592 Enamine

2-hydrazinylbenzene-1-sulfonamide

  • MDL number MFCD03093797
  • Cas Number : 90824-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185937

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185937
Availability details for
Quantity
  • Availability
  • Add to Cart