ENA428683606 Enamine

2-[3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl]-2,3-dihydro-1H-indole

  • MDL number MFCD13718603
  • Cas Number : 1154346-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185964
Availability details for
Quantity
  • Availability
  • Add to Cart