ENA428683607 Enamine

2-(3-fluoro-4-methoxyphenyl)-2-oxoethyl 6-cyclopropyl-3-methyl-[1,2]oxazolo[5,4-b]pyridine-4-carboxylate

  • MDL number MFCD28118555
  • Cas Number : 1061175-49-2
  • Molecular Weight : 384.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-185965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185965
Availability details for
Quantity
  • Availability
  • Add to Cart