ENA428683612 Enamine

1-[4-hydroxy-3-(propan-2-yl)phenyl]-2-methylpropan-1-one

  • MDL number MFCD24076352
  • Cas Number : 1508439-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185984
Availability details for
Quantity
  • Availability
  • Add to Cart