ENA428683616 Enamine

3-amino-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD20719838
  • Cas Number : 15908-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185532
Availability details for
Quantity
  • Availability
  • Add to Cart