ENA428683617 Enamine

N-methyl-6-(trifluoromethyl)pyridazin-3-amine

  • MDL number MFCD13705388
  • Cas Number : 1216271-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-152
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185998

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185998
Availability details for
Quantity
  • Availability
  • Add to Cart