ENA428683619 Enamine

tert-butyl N-{[3-amino-5-(trifluoromethyl)phenyl]methyl}carbamate

  • MDL number MFCD24470488
  • Cas Number : 693249-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186762
Availability details for
Quantity
  • Availability
  • Add to Cart