ENA428683628 Enamine

cyclohexyl 2-sulfanylacetate

  • MDL number MFCD00046348
  • Cas Number : 16849-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186774

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186774
Availability details for
Quantity
  • Availability
  • Add to Cart