ENA428683631 Enamine

6-(2-fluorophenyl)pyrimidine-2,4-diamine

  • MDL number MFCD16103643
  • Cas Number : 363-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185564

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185564
Availability details for
Quantity
  • Availability
  • Add to Cart