ENA428683634 Enamine

4-bromo-1-(2-methanesulfonylethyl)-3,5-dimethyl-1H-pyrazole

  • MDL number MFCD16691240
  • Cas Number : 1247899-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186804
Availability details for
Quantity
  • Availability
  • Add to Cart