ENA428683637 Enamine

N-(2-phenylethyl)benzenesulfonamide

  • MDL number MFCD00963360
  • Cas Number : 77198-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186807

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186807
Availability details for
Quantity
  • Availability
  • Add to Cart