ENA428683645 Enamine

2-(3-methyl-1,2,4-oxadiazol-5-yl)-5-(trifluoromethyl)aniline

  • MDL number MFCD22702328
  • Cas Number : 1503695-84-8
  • Molecular Weight : 243.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-186841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186841
Availability details for
Quantity
  • Availability
  • Add to Cart