ENA428683646 Enamine

4-(difluoromethoxy)-5-fluoro-2-(1H-1,2,4-triazol-1-yl)aniline

  • MDL number MFCD18986285
  • Cas Number : 1411217-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186842

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186842
Availability details for
Quantity
  • Availability
  • Add to Cart