ENA428683648 Enamine

(1r,3r)-3-phenylcyclobutan-1-ol

  • MDL number MFCD22395419
  • Cas Number : 150639-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185580

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-185580
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart